(S)-(-)-α-Methylbenzylamin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(S)-(-)-1-Phenylethylamin
P0793-25ML
P0793-500ML
P0793-100ML
TCIAP0793-25MLEA
263
DKK
TCIAP0793-25ML
TCIAP0793-500ML
TCIAP0793-100ML
(S)-(-)-α-Methylbenzylamin ≥98.0% (by GC)
(S)-(-)-α-Methylbenzylamin
Formel:
C₈H₁₁N Molekylvægt: 121.18 g/mol Kogepunkt: 84°C Smeltepunkt: -10 Flammepunkt: 79°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064406 CAS-nummer: 2627-86-3 EINECS: 220-098-0 UN nummer: 2922 ADR: 8 II Merck Index: 12,06106 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific gravity (20/20) | 0.9510 to 0.9560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...