N,N,N',N'-Tetramethyl-p-phenylendiamin dihydrochlorid ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride
T0151-1G
T0151-25G
T0151-5G
TCIAT0151-1GEA
132
DKK
TCIAT0151-1G
TCIAT0151-25G
TCIAT0151-5G
N,N,N',N'-Tetramethyl-p-phenylendiamin dihydrochlorid ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetramethyl-p-phenylendiamin dihydrochlorid
Formel:
C₁₀H₁₈Cl₂N₂ Molekylvægt: 237.17 g/mol Smeltepunkt: 224°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012482 CAS-nummer: 637-01-4 EINECS: 211-274-8 Merck Index: 12,09367 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...