(±)-1,2-Decandiol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D2720-25G
D2720-100G
TCIAD2720-25GEA
288
DKK
TCIAD2720-25G
TCIAD2720-100G
(±)-1,2-Decandiol ≥99.0% (by GC)
(±)-1,2-Decandiol
Formel:
C₁₀H₂₂O₂ Kogepunkt: 168°C |
MDL Number:
MFCD00010739 CAS-nummer: 1119-86-4 EINECS: 214-288-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 47.0 to 50.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...