(±)-3-Chlor-1,2-propandiol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (±)-3-Chloro-1,2-dihydroxy propane

C0268-25G C0268-500G
TCIAC0268-25GEA 132 DKK
TCIAC0268-25G TCIAC0268-500G
(±)-3-Chlor-1,2-propandiol ≥98.0% (by GC)
(±)-3-Chlor-1,2-propandiol
Formel: C₃H₇ClO₂
Molekylvægt: 110.54 g/mol
Kogepunkt: 213
Smeltepunkt: –40°C
Flammepunkt: 137°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004712
CAS-nummer: 96-24-2
EINECS: 202-492-4
UN nummer: 2689
Merck Index: 14,02145
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b5b9c0d4c82075b9a1581610a1478636706350d&vid=9b5b9c0d4c82075b9a1581610a1478636706350d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR