Zinkchlorid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
Z0014-25G
Z0014-300G
TCIAZ0014-25GEA
165
DKK
TCIAZ0014-25G
TCIAZ0014-300G
Zinkchlorid ≥98.0% (by titrimetric analysis)
Zinkchlorid
Formel:
ZnCl₂ Molekylvægt: 136.3 g/mol Kogepunkt: 732°C Smeltepunkt: 290°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011295 CAS-nummer: 7646-85-7 EINECS: 231-592-0 UN nummer: 2331 ADR: 8 III Merck Index: 14,10132 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...