trans-4-Methoxycinnamic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: trans-O-Methyl-p-coumaric Acid, (E)-3-(4-Methoxyphenyl)-2-propenoic acid

M0576-25G M0576-100G
TCIAM0576-25GEA 279 DKK
TCIAM0576-25G TCIAM0576-100G
trans-4-Methoxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Methoxycinnamic acid
Formel: C₁₀H₁₀O₃
MDL Number: MFCD00004398
CAS-nummer: 943-89-5
EINECS: 213-405-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43c48df52f2bffb481ca59e9f705950754e4472b&vid=9033cfdf933b57b38bda633e02e6f02258b68573&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Mesomorphic range 168.0 to 193.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR