4-(Trifluormethyl)kanelsyre ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: trans-4-(Trifluoromethyl)cinnamic acid

T2499-1G T2499-25G T2499-5G
TCIAT2499-1GEA 156 DKK
TCIAT2499-1G TCIAT2499-25G TCIAT2499-5G
4-(Trifluormethyl)kanelsyre ≥98.0% (by GC)
4-(Trifluormethyl)kanelsyre
Formel: C₁₀H₇F₃O₂
Molekylvægt: 216.16 g/mol
MDL Number: MFCD00002696
CAS-nummer: 16642-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1a83b9fb1777b29cb28ce9137b5dcc4e1c8e49a&vid=c1a83b9fb1777b29cb28ce9137b5dcc4e1c8e49a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR