Dimethylfumarat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Fumaric acid dimethyl ester

F0069-25G F0069-500G
TCIAF0069-25GEA 140 DKK
TCIAF0069-25G TCIAF0069-500G
Dimethylfumarat ≥98.0% (by GC)
Dimethylfumarat
Formel: C₆H₈O₄
Kogepunkt: 205°C
Smeltepunkt: 102 °C
Massefylde: 1,37 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064438
CAS-nummer: 624-49-7
EINECS: 210-849-0
Merck Index: 14,04287
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=347c694ca807cb29d6a611fb07b55460e096b661&vid=347c694ca807cb29d6a611fb07b55460e096b661&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR