Tetramethylammoniumchlorid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0136-25G T0136-500G
TCIAT0136-25GEA 140 DKK
TCIAT0136-25G TCIAT0136-500G
Tetramethylammoniumchlorid ≥98.0% (by titrimetric analysis)
Tetramethylammonium chloride
Formel: (CH₃)₄NCl
Smeltepunkt: > 300 °C
Massefylde: 1,169 g/cm³ (20 °C)
Flammepunkt: ≥ 100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011628
CAS-nummer: 75-57-0
EINECS: 200-880-8
UN nummer: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0abdb045a6d68f186ae980c7f9df12ad066c5e4&vid=a0abdb045a6d68f186ae980c7f9df12ad066c5e4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR