Polyethylen glycol 6.000
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
PEG 6,000, PEG 6000
P0903-25G
P0903-500G
TCIAP0903-25GEA
132
DKK
TCIAP0903-25G
TCIAP0903-500G
Polyethylen glycol 6.000
Polyethylen glycol 6.000
Formel:
H(OCH₂CH₂)nOH Molekylvægt: 6000 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00081839 CAS-nummer: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,07568 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Melting point | 62.0 to 65.0 deg-C |
Solubility in Water | almost transparency |
pH | 5.0 to 8.0 (50 g/L, 25 deg-C) |
Average molecular weight | 7,300 to 9,300 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...