Polyethylen glycol 600

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethylene glycol polymer average Mn 600, PEG 600

P1187-25G P1187-500G
TCIAP1187-25GEA 132 DKK
TCIAP1187-25G TCIAP1187-500G
Polyethylen glycol 600
Polyethylen glycol 600
Formel: H(OCH₂CH₂)nOH
Molekylvægt: 600 g/mol
Flammepunkt: 270
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081839
CAS-nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,07568

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Freezing point 17.0 to 21.0 deg-C
pH 4.0 to 7.0 (50g/L, 25deg-c)
Average molecular weight 550 to 650


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR