Valeryl chlorid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Pentanoyl chloride, Valeroyl chloride
V0010-25ML
V0010-500ML
V0010-100ML
TCIAV0010-25MLEA
132
DKK
TCIAV0010-25ML
TCIAV0010-500ML
TCIAV0010-100ML
Valeryl chlorid ≥98.0% (by titrimetric analysis)
Valeryl chlorid
Formel:
C₅H₉ClO Kogepunkt: 126°C Smeltepunkt: –110°C Massefylde: 1,016 g/cm³ (20 °C) Flammepunkt: 32°C |
MDL Number:
MFCD00000757 CAS-nummer: 638-29-9 EINECS: 211-330-1 UN nummer: 2502 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...