Trimethylborat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B0522-25ML B0522-500ML B0522-100ML
TCIAB0522-25MLEA 140 DKK
TCIAB0522-25ML TCIAB0522-500ML TCIAB0522-100ML
Trimethylborat ≥98.0% (by titrimetric analysis)
Trimethylborat
Formel: C₃H₉BO₃
Kogepunkt: 67 °C (1013 hPa)
Smeltepunkt: -29°C
Massefylde: 0,915 g/cm³ (20 °C)
Flammepunkt: -8°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00008346
CAS-nummer: 121-43-7
EINECS: 204-468-9
UN nummer: 2416
ADR: 3,II
Merck Index: 14,09712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ead84429d4015e754172931f5028eb4745100a8&vid=0ead84429d4015e754172931f5028eb4745100a8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 98.0 %
Boric acid max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR