N,N-Diethylformamid ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Formic acid diethylamide, Formdiethylamide

D0506-25ML D0506-500ML D0506-100ML
TCIAD0506-25MLEA 173 DKK
TCIAD0506-25ML TCIAD0506-500ML TCIAD0506-100ML
N,N-Diethylformamid ≥99.0% (by GC)
N,N-Diethylformamid
Formel: C₅H₁₁NO
Kogepunkt: 177°C
Massefylde: 0,908 g/cm³ (20 °C)
Flammepunkt: 60°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003287
CAS-nummer: 617-84-5
EINECS: 210-533-2
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b585ec202c948282af306bb85baaddf12a0385d&vid=4b585ec202c948282af306bb85baaddf12a0385d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR