Ethyl methyl sulfid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

E0143-100ML E0143-25ML
TCIAE0143-100MLEA 575 DKK
TCIAE0143-100ML TCIAE0143-25ML
Ethyl methyl sulfid ≥98.0% (by GC)
Ethyl methyl sulfid
Formel: C₃H₈S
Molekylvægt: 76.16 g/mol
Kogepunkt: 66°C
Smeltepunkt: -104.8
Flammepunkt: –15°C
MDL Number: MFCD00009268
CAS-nummer: 624-89-5
EINECS: 210-868-4
UN nummer: 1993
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45be1bc827a1233cfbff9a70179aa3b26dda38ce&vid=45be1bc827a1233cfbff9a70179aa3b26dda38ce&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR