Tetramethylthiurammonosulfid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(dimethylthiocarbamyl) sulphide

T0157-25G T0157-500G
TCIAT0157-25GEA 132 DKK
TCIAT0157-25G TCIAT0157-500G
Tetramethylthiurammonosulfid ≥98.0% (by GC)
Tetramethylthiurammonosulfid
Formel: C₆H₁₂N₂S₃
Molekylvægt: 208.37 g/mol
Smeltepunkt: 110
MDL Number: MFCD00014870
CAS-nummer: 97-74-5
EINECS: 202-605-7
UN nummer: 3077
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed63a98b8d44c380a166c098b952d4180896a1df&vid=ed63a98b8d44c380a166c098b952d4180896a1df&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 108.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR