Trichloracetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0372-25G T0372-500G T0372-100G
TCIAT0372-25GEA 156 DKK
TCIAT0372-25G TCIAT0372-500G TCIAT0372-100G
Trichloracetonitril ≥98.0% (by GC)
Trichloracetonitril
Formel: C₂Cl₃N
Kogepunkt: 85°C
Smeltepunkt: –44 °C
Massefylde: 1,44 g/cm³ (20 °C)
Flammepunkt: 195°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001842
CAS-nummer: 545-06-2
EINECS: 208-885-7
UN nummer: 3276
ADR: 6.1,III
Merck Index: 14,09628
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d90316842060ef5e274bbdcc0fb2021e2eac96d&vid=7d90316842060ef5e274bbdcc0fb2021e2eac96d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR