Chlortrimethylsilan ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: TMCS, TMSC, Trimethylchlorosilane
C0306-25ML C0306-500ML C0306-100ML
TCIAC0306-25MLEA 140 DKK
TCIAC0306-25ML TCIAC0306-500ML TCIAC0306-100ML
Chlortrimethylsilan ≥98.0% (by GC)
Chlortrimethylsilan
Formel: C₃H₉ClSi
Kogepunkt: 57°C
Smeltepunkt: -58°C
Massefylde: 0,8576 g/cm³ (20 °C)
Flammepunkt: -28°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000502
CAS-nummer: 75-77-4
EINECS: 200-900-5
UN nummer: 1298
ADR: 3,II
REACH: 01-2119457596-25
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609bf473b5db5c5490eae6a445b03453c76c1ad2&vid=609bf473b5db5c5490eae6a445b03453c76c1ad2&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %