Pyrrolidin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Tetrahydropyrrole
P0576-25ML
P0576-500ML
TCIAP0576-25MLEA
132
DKK
TCIAP0576-25ML
TCIAP0576-500ML
Pyrrolidin ≥98.0% (by GC)
Pyrrolidin
Formel:
C₄H₉N Kogepunkt: 88°C Smeltepunkt: –63°C Massefylde: 0,86 g/cm³ (20 °C) Flammepunkt: 3°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005249 CAS-nummer: 123-75-1 EINECS: 204-648-7 UN nummer: 1922 ADR: 3,II Merck Index: 14,08015 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...