7-Chlorindol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2436-1G C2436-5G
TCIAC2436-1GEA 444 DKK
TCIAC2436-1G TCIAC2436-5G
7-Chlorindol ≥98.0% (by GC)
7-Chloroindole
Formel: C₈H₆ClN
MDL Number: MFCD01321301
CAS-nummer: 53924-05-3
EINECS: 258-865-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35220f55ec4bcb32cb4dcdcbce9985c52fd95061&vid=35220f55ec4bcb32cb4dcdcbce9985c52fd95061&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR