Indol-6-carboxylsyre ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 1H-Indole-6-carboxylic acid
I0663-1G I0663-5G
TCIAI0663-1GEA 263 DKK
TCIAI0663-1G TCIAI0663-5G
Indol-6-carboxylsyre ≥98.0% (by HPLC, titration analysis)
Indol-6-carboxylsyre
Formel: C₉H₇NO₂
Molekylvægt: 161.16 g/mol
Smeltepunkt: 257°C
MDL Number: MFCD00210441
CAS-nummer: 1670-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76a69f9505b7b1a4da88f64de9bddea0c65a913f&vid=76a69f9505b7b1a4da88f64de9bddea0c65a913f&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %