Indol-6-carbonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
6-Cyano-1H-indole, 6-Cyanoindole, 1H-Indole-6-carbonitrile
C2084-25G
C2084-5G
TCIAC2084-25GEA
3300
DKK
TCIAC2084-25G
TCIAC2084-5G
Indol-6-carbonitril ≥98.0% (by GC)
Indol-6-carbonitril
Formel:
C₉H₆N₂ Molekylvægt: 142.16 g/mol |
MDL Number:
MFCD00016732 CAS-nummer: 15861-36-6 EINECS: 239-988-5 UN nummer: 3439 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...