Bromcresol lilla ≥97.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B0580-1G
B0580-25G
TCIAB0580-1GEA
181
DKK
TCIAB0580-1G
TCIAB0580-25G
Bromcresol lilla ≥97.0% (by HPLC)
Bromcresol lilla
Formel:
C₂₁H₁₆Br₂O₅S Smeltepunkt: 242 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011681 CAS-nummer: 115-40-2 EINECS: 204-087-8 Merck Index: 14,01387 |
|
Specifikationer Testresultater
Appearance | White to Brown to Dark purple powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Absorbance | min. 0.540 (5mg/L, pH 7.2 buffer sol., 588.0 to 592.0 nm) |
Solubility in EtOH (50vol%) | almost transparency |
Transition interval (pH) | (yellow)5.2 to 6.8 (purple) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...