4-Methylpiperidin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
P0445-25ML
P0445-500ML
TCIAP0445-25MLEA
247
DKK
TCIAP0445-25ML
TCIAP0445-500ML
4-Methylpiperidin ≥98.0% (by GC, titration analysis)
4-Methylpiperidin
Formel:
C₆H₁₃N Kogepunkt: 127°C Smeltepunkt: –4 °C Massefylde: 0,84 g/cm³ (20 °C) Flammepunkt: 9°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006005 CAS-nummer: 626-58-4 EINECS: 210-954-1 UN nummer: 1993 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...