Butyl 4-hydroxybenzoat ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

H0210-25G H0210-500G
TCIAH0210-25GEA 132 DKK
TCIAH0210-25G TCIAH0210-500G
Butyl 4-hydroxybenzoat ≥99.0% (by GC, titration analysis)
Butyl 4-hydroxybenzoat
Formel: C₁₁H₁₄O₃
Kogepunkt: 156°C
Smeltepunkt: 73 °C
Massefylde: 1,23 g/cm³ (20 °C)
MDL Number: MFCD00016478
CAS-nummer: 94-26-8
EINECS: 202-318-7
Merck Index: 14,01584
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96a4c0cb50a7841b108affb19507af3b287f209&vid=a96a4c0cb50a7841b108affb19507af3b287f209&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 69.0 to 72.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR