4-fluorbenzamid ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

F0279-25G F0279-5G
TCIAF0279-25GEA 1080 DKK
TCIAF0279-25G TCIAF0279-5G
4-fluorbenzamid ≥98.0% (by total nitrogen basis)
4-fluorbenzamid
Formel: C₇H₆FNO
Molekylvægt: 139.13 g/mol
Kogepunkt: 287
Smeltepunkt: 155
MDL Number: MFCD00007992
CAS-nummer: 824-75-9
EINECS: 212-535-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4780cb9a917bec9c2dfb6157fa68158e4d3f51d&vid=a4780cb9a917bec9c2dfb6157fa68158e4d3f51d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 154.0 to 157.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR