4-Brom-2-nitrobenzoesyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
B3810-25G B3810-5G
TCIAB3810-25GEA 1210 DKK
TCIAB3810-25G TCIAB3810-5G
4-Brom-2-nitrobenzoesyre ≥98.0% (by titrimetric analysis)
4-Brom-2-nitrobenzoesyre
Formel: C₇H₄BrNO₄
MDL Number: MFCD01013599
CAS-nummer: 99277-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=294c7b54343cc6fc3a08810531dcca51f0d1c906&vid=294c7b54343cc6fc3a08810531dcca51f0d1c906&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C