4-Aminobutan-1-ol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A1013-25G A1013-5G
TCIAA1013-25GEA 1790 DKK
TCIAA1013-25G TCIAA1013-5G
4-Aminobutan-1-ol ≥98.0% (by GC)
4-Aminobutan-1-ol
Formel: C₄H₁₁NO
Kogepunkt: 206°C
Smeltepunkt: 16…18 °C
Massefylde: 0,967 g/cm³ (20 °C)
Flammepunkt: 108°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008230
CAS-nummer: 13325-10-5
EINECS: 236-364-4
UN nummer: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8210b631a54dec476b9bf4e1d7b8c551f3e36ad1&vid=8210b631a54dec476b9bf4e1d7b8c551f3e36ad1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR