3-Methyl-3-pentanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M0387-25ML M0387-100ML
TCIAM0387-25MLEA 362 DKK
TCIAM0387-25ML TCIAM0387-100ML
3-Methyl-3-pentanol ≥98.0% (by GC)
3-Methyl-3-pentanol
Formel: C₆H₁₄O
Kogepunkt: 126°C
Smeltepunkt: –53 °C
Massefylde: 0,8452 g/cm³ (0 °C)
Flammepunkt: 24°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004483
CAS-nummer: 77-74-7
EINECS: 201-053-4
UN nummer: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f45af6dc4662456d35b01d86cd06e5035f5481cf&vid=f45af6dc4662456d35b01d86cd06e5035f5481cf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR