3-Methyl-1-butanol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

I0289-25ML I0289-500ML
TCIAI0289-25MLEA 156 DKK
TCIAI0289-25ML TCIAI0289-500ML
3-Methyl-1-butanol ≥99.0% (by GC)
Isoamyl alcohol
Formel: (CH₃)₂CHCH₂CH₂OH
Kogepunkt: 132°C
Smeltepunkt: –117 °C
Massefylde: 0,812 g/cm³ (20 °C)
Flammepunkt: 46°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002934
CAS-nummer: 123-51-3
EINECS: 204-633-5
UN nummer: 1105
ADR: 3,III
Merck Index: 14,05195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f84b5fac2d9d87ddc43aef8ed62f60d80c864d8&vid=4f84b5fac2d9d87ddc43aef8ed62f60d80c864d8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR