Methyl3-mercaptopropionat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Mercaptopropionic acid methyl ester, Methyl 3-mercaptopropanoate

M1434-25G M1434-500G
TCIAM1434-25GEA 345 DKK
TCIAM1434-25G TCIAM1434-500G
Methyl3-mercaptopropionat ≥98.0% (by GC)
Methyl3-mercaptopropionat
Formel: C₄H₈O₂S
Kogepunkt: 54 °C (1013 hPa)
Smeltepunkt: –60 °C
Massefylde: 1,085 g/cm³ (20 °C)
Flammepunkt: 64°C
MDL Number: MFCD00004895
CAS-nummer: 2935-90-2
EINECS: 220-912-4
UN nummer: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8aa77197e413a2d8504cf31235bf0fe170dc8968&vid=8aa77197e413a2d8504cf31235bf0fe170dc8968&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR