3-Chlor-2,4,5,6-tetrafluorpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Chlorotetrafluoropyridine
C1549-25G
C1549-5G
TCIAC1549-25GEA
887
DKK
TCIAC1549-25G
TCIAC1549-5G
3-Chlor-2,4,5,6-tetrafluorpyridin ≥98.0% (by GC)
3-Chlor-2,4,5,6-tetrafluorpyridin
Formel:
C₅ClF₄N Molekylvægt: 185.51 g/mol Kogepunkt: 71°C |
MDL Number:
MFCD00010624 CAS-nummer: 1735-84-8 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...