5-Cloro-2-nitrobenzamid ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C1978-25G
C1978-5G
TCIAC1978-25GEA
3850
DKK
TCIAC1978-25G
TCIAC1978-5G
5-Cloro-2-nitrobenzamid ≥90.0% (by GC)
5-Cloro-2-nitrobenzamid
Formel:
C₇H₅ClN₂O₃ Molekylvægt: 200.58 g/mol |
MDL Number:
MFCD00017509 CAS-nummer: 40763-96-0 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...