5-Brom-2-fluorpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B2705-25G
B2705-5G
TCIAB2705-25GEA
928
DKK
TCIAB2705-25G
TCIAB2705-5G
5-Brom-2-fluorpyridin ≥98.0% (by GC)
5-Brom-2-fluorpyridin
Formel:
C₅H₃BrFN Kogepunkt: 63°C Flammepunkt: 73°C |
MDL Number:
MFCD01863742 CAS-nummer: 766-11-0 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...