5-Amino-2-chlorpyridin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A1155-1G A1155-5G
TCIAA1155-1GEA 222 DKK
TCIAA1155-1G TCIAA1155-5G
5-Amino-2-chlorpyridin ≥98.0% (by GC, titration analysis)
5-Amino-2-chlorpyridin
Formel: C₅H₅ClN₂
Kogepunkt: 225 °C (1013 hPa)
Smeltepunkt: 83 °C
MDL Number: MFCD00006243
CAS-nummer: 5350-93-6
EINECS: 226-322-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e47c157992c582e09b03eab8f5245a79f21649e&vid=9e47c157992c582e09b03eab8f5245a79f21649e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR