3-Amino-p-benzoesyre ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A1955-25G
A1955-5G
TCIAA1955-25GEA
2950
DKK
TCIAA1955-25G
TCIAA1955-5G
3-Amino-p-benzoesyre ≥98.0% (by HPLC, titration analysis)
3-Amino-p-benzoesyre
Formel:
C₈H₉NO₃ Molekylvægt: 167.16 g/mol Kogepunkt: 335 Smeltepunkt: 204 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002521 CAS-nummer: 2840-26-8 EINECS: 220-634-3 |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 203.0 to 209.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...