3,5-Dichlor-2-fluorpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D3317-1G
D3317-5G
TCIAD3317-1GEA
132
DKK
TCIAD3317-1G
TCIAD3317-5G
3,5-Dichlor-2-fluorpyridin ≥98.0% (by GC)
Formel:
C₅H₂Cl₂FN |
MDL Number:
MFCD04039349 CAS-nummer: 823-56-3 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...