3,3-Dimethylsmørsyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3,3-Dimethylbutanoic acid, tert-Butylacetic acid

B0826-25ML B0826-500ML
TCIAB0826-25MLEA 255 DKK
TCIAB0826-25ML TCIAB0826-500ML
3,3-Dimethylsmørsyre ≥98.0% (by GC, titration analysis)
3,3-Dimethylsmørsyre
Formel: C₆H₁₂O₂
Molekylvægt: 116.16 g/mol
Kogepunkt: 185
Smeltepunkt: 7
Massefylde: 0.9303
Flammepunkt: 89°C
MDL Number: MFCD00002715
CAS-nummer: 1070-83-3
EINECS: 213-982-2
UN nummer: 3265
ADR: 8 III
Merck Index: 14,01538
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c47b41a03f91d4fcecbb9de6cd543861002c50dc&vid=c47b41a03f91d4fcecbb9de6cd543861002c50dc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR