2-Methyl-6-phenylpyridin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M2001-1G M2001-5G
TCIAM2001-1GEA 1020 DKK
TCIAM2001-1G TCIAM2001-5G
2-Methyl-6-phenylpyridin ≥98.0% (by GC, titration analysis)
Formel: C₁₂H₁₁N
Kogepunkt: 155°C
MDL Number: MFCD00796537
CAS-nummer: 46181-30-0
EINECS: 256-258-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de9fffbae6ec9d7c474d3c4eaa8331e65b7149a1&vid=de9fffbae6ec9d7c474d3c4eaa8331e65b7149a1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR