2-Fluor-5-methylbenzoesyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0426-25G F0426-5G F0426-1G
TCIAF0426-25GEA 1690 DKK
TCIAF0426-25G TCIAF0426-5G TCIAF0426-1G
2-Fluor-5-methylbenzoesyre ≥98.0% (by titrimetric analysis)
2-Fluor-5-methylbenzoesyre
Formel: C₈H₇FO₂
MDL Number: MFCD00092819
CAS-nummer: 321-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1570ea6bd7247d91846d46f02705fa64d5cce90&vid=d1570ea6bd7247d91846d46f02705fa64d5cce90&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 159.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR