2-Chlor-5-fluorbenzoesyre ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2088-25G
C2088-5G
TCIAC2088-25GEA
879
DKK
TCIAC2088-25G
TCIAC2088-5G
2-Chlor-5-fluorbenzoesyre ≥98.0% (by GC)
2-Chlor-5-fluorbenzoesyre
Formel:
C₇H₄ClFO₂ |
MDL Number:
MFCD00156967 CAS-nummer: 2252-50-8 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...