2-Bromethanol ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B0590-25G
B0590-500G
B0590-100G
TCIAB0590-25GEA
132
DKK
TCIAB0590-25G
TCIAB0590-500G
TCIAB0590-100G
2-Bromethanol ≥95.0% (by GC)
2-Bromoethanol
Formel:
BrH₂CCH₂OH Kogepunkt: 149 °C (1013 hPa) Smeltepunkt: –80 °C Massefylde: 1,76 g/cm³ (20 °C) Flammepunkt: 40 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00002827 CAS-nummer: 540-51-2 EINECS: 208-748-1 UN nummer: 2922 ADR: 8,II Merck Index: 14,03792 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...