2-Amino-5-methylpyridin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline
A0732-25G
A0732-500G
TCIAA0732-25GEA
304
DKK
TCIAA0732-25G
TCIAA0732-500G
2-Amino-5-methylpyridin ≥98.0% (by GC, titration analysis)
2-Amino-5-methylpyridine
Formel:
C₆H₈N₂ Molekylvægt: 108.14 g/mol Kogepunkt: 227.1 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006328 CAS-nummer: 1603-41-4 EINECS: 216-503-5 UN nummer: 2811 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 76.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...