6-Amino-3-Brom-2-methylpyridin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A1889-1G A1889-5G
TCIAA1889-1GEA 148 DKK
TCIAA1889-1G TCIAA1889-5G
6-Amino-3-Brom-2-methylpyridin ≥98.0% (by GC, titration analysis)
6-Amino-3-Brom-2-methylpyridin
Formel: C₆H₇BrN₂
MDL Number: MFCD00068230
CAS-nummer: 42753-71-9
EINECS: 255-927-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21c0d386ff3b8e3f14386476b7cb78710ddf77fa&vid=21c0d386ff3b8e3f14386476b7cb78710ddf77fa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR