1-Methylpyrrolidin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M0415-25ML
M0415-500ML
M0415-100ML
TCIAM0415-25MLEA
140
DKK
TCIAM0415-25ML
TCIAM0415-500ML
TCIAM0415-100ML
1-Methylpyrrolidin ≥98.0% (by GC, titration analysis)
1-Methylpyrrolidin
Formel:
C₅H₁₁N Kogepunkt: 80 °C (1013 hPa) Smeltepunkt: –76 °C Massefylde: 0,799 g/cm³ (20 °C) Flammepunkt: 3°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003173 CAS-nummer: 120-94-5 EINECS: 204-438-5 UN nummer: 2924 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...