o-Cresol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
o-Methylphenol
C0402-25G
C0402-500G
TCIAC0402-25GEA
140
DKK
TCIAC0402-25G
TCIAC0402-500G
o-Cresol ≥99.0% (by GC)
o-Cresol
Formel:
C₇H₈O Molekylvægt: 108.14 g/mol Kogepunkt: 192°C Flammepunkt: 81 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002226 CAS-nummer: 95-48-7 EINECS: 202-423-8 UN nummer: 3455 ADR: 6.1 II Merck Index: 14,02579 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Solubility in Sodium hydroxide solution | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...