2-Fluorbenzylalkohol ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2-Fluorophenyl)methanol, 2-Fluorobenzyl Alcohol

F0259-25G F0259-5G
TCIAF0259-25GEA 591 DKK
TCIAF0259-25G TCIAF0259-5G
2-Fluorbenzylalkohol ≥97.0% (by GC)
2-Fluorbenzylalkohol
Formel: C₇H₇FO
Kogepunkt: 96°C
Massefylde: 1,17 g/cm³ (20 °C)
Flammepunkt: 90°C
MDL Number: MFCD00004601
CAS-nummer: 446-51-5
EINECS: 207-170-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a46bbb8274b06ab28652426ff073b3e72c6b5ac&vid=3a46bbb8274b06ab28652426ff073b3e72c6b5ac&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR