3'-Methoxyacetophenon ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M0104-25ML M0104-500ML
TCIAM0104-25MLEA 140 DKK
TCIAM0104-25ML TCIAM0104-500ML
3'-Methoxyacetophenon ≥97.0% (by GC)
3'-Methoxyacetophenon
Formel: C₉H₁₀O₂
Kogepunkt: 240°C
Smeltepunkt: –8 °C
Massefylde: 1,09 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008736
CAS-nummer: 586-37-8
EINECS: 209-573-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f89c18f9199c2c0be2ad2bbe033867bb632b7a29&vid=f89c18f9199c2c0be2ad2bbe033867bb632b7a29&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR