3,4,5-Trifluorbenzonitril ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T2370-1G T2370-5G
TCIAT2370-1GEA 321 DKK
TCIAT2370-1G TCIAT2370-5G
3,4,5-Trifluorbenzonitril ≥98.0% (by GC, total nitrogen)
3,4,5-Trifluorbenzonitril
Formel: C₇H₂F₃N
Kogepunkt: 69°C
Smeltepunkt: 45…47 °C
MDL Number: MFCD00074948
CAS-nummer: 134227-45-5
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6018752c6f900ec809c871f7971bb1e8624fd578&vid=6018752c6f900ec809c871f7971bb1e8624fd578&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR