4-Chlor-o-cresol ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-Chloro-4-hydroxy-3-methylbenzene, 4-Chloro-2-methylphenol

C0152-25G C0152-500G
TCIAC0152-25GEA 238 DKK
TCIAC0152-25G TCIAC0152-500G
4-Chlor-o-cresol ≥90.0% (by GC)
4-Chlor-o-cresol
Formel: C₇H₇ClO
Molekylvægt: 142.58 g/mol
Kogepunkt: 224
Flammepunkt: 78
MDL Number: MFCD00002321
CAS-nummer: 1570-64-5
EINECS: 216-381-3
UN nummer: 3437
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98aefc05bc5bbde31fa79ca537cb08504896671c&vid=98aefc05bc5bbde31fa79ca537cb08504896671c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR